Learn More
Sino Biological Peptide Substrates: Tyr-phosphopeptide-2
Supplier:  Sino Biological T7058
Encompass_Preferred
The Tyr-phosphopeptide-2 sub peptide sequence (DADEY(p)LIPDQG) is based on mouse epidermal growth factor receptor isoform 1 (amino acid 1014-1024).
Catalog No. 50-256-7583