Learn More
CPC Scientific H-Tyr-Glu-Ala-Glu-Asp-Leu-Gln-Val-Gly-Gln-Val-Glu-Leu-Gly-Gly-Gly-Pro-Gly-Ala-Gly-Ser-Leu-Gln-Pro-Leu-Ala-Leu-Glu-Gly-Ser-Leu-Gln-OH (acetate salt) 0.5MG
SDP
Supplier:  CPC Scientific CPEP002A
Encompass
ONE-LETTER SEQUENCE: YEAEDLQVGQVELGGGPGAGSLQPLALEGSLQMOLECULAR FORMULA: C138H220N36O50MOLECULAR WEIGHT:3183.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [57327-90-9], RESEARCH AREA: DiabetesREFERENCES: V.K. Naithani et al., Hoppe Seylers Z. Physiol. Chem., 356, 1305 (1975); N. Yanaihara et al., Hoppe Seylers Z. Physiol. Chem., 362, 775 (1981); H. Sun et al., Appl. Biochem. Biotechnol., 55, 167 (1995)
Catalog No. 50-210-9149