Learn More
CPC Scientific H-Tyr-Ala-Cys-Asn-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Met-Val-Lys-Ser-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Lys-Ala-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP022B
Encompass
ONE-LETTER SEQUENCE: YACNTATCVTHRLAGLLSRSGGMVKSNFVPTNVGSKAF-NH2 (Cys3 and 8 bridge)MOLECULAR FORMULA: C171H276N52O50S3MOLECULAR WEIGHT:3956.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: Cardiovascular
Catalog No. 50-211-0040