Learn More
CPC Scientific H-Tyr-Ser-Cys-Asn-Thr-Ala-Thr-Cys-Val-Thr-His-Arg-Leu-Ala-Gly-Leu-Leu-Ser-Arg-Ser-Gly-Gly-Val-Val-Lys-Asp-Asn-Phe-Val-Pro-Thr-Asn-Val-Gly-Ser-Glu-Ala-Phe-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OSTP024B
Encompass
ONE-LETTER SEQUENCE: YSCNTATCVTHRLAGLLSRSGGVVKDNFVPTNVGSEAF-NH2 (Cys3 and 8 bridge)MOLECULAR FORMULA: C171H271N51O54S2MOLECULAR WEIGHT:3969.5STORAGE CONDITIONS: -20 5CSYNONYMS: Tyr-alpha-CGRP (rat), Tyr-CGRP I (rat)RESEARCH AREA: Cardiovascular
Catalog No. 50-211-0044