Learn More
Medchemexpress LLC Tyr3-octreotate | 302794-43-0 | 99.76% | 1049.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP102391MG
Encompass_Preferred
Tyr3-Octreotate is a somatostatin analog that exhibits high uptake into tumors. It can be labeled with radioactive metal, demonstrating antitumor efficacy. This product can be used for the synthesis/research of Radionuclide-Drug Conjugates (RDCs).
Catalog No. 50-004-25279