Learn More
Medchemexpress LLC Tyramine hydrochloride | 60-19-5 | MFCD00012901 | 99.8% | 173.64 | C8H12ClNO | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0168231G
Encompass_Preferred
Tyramine hydrochloride is the hydrochloride salt of the endogenous monoamine tyramine, supplied as a solid reference standard for biochemical and neuroscience research. It is used in studies of monoamine metabolism and neuromodulation, and as an analytical standard for chromatography and mass spectrometry.
  • High purity (99.82%) suitable for analytical and research applications.
  • White to light yellow solid, stable for storage and handling.
  • Chemical formula C8H12ClNO and molecular weight 173.64 g/mol.
  • Intended for use in neuroscience and neuromodulation studies.
  • Available in multiple package sizes to support small-scale experiments.
Catalog No. 50-003-00421