Learn More
TYRELL MEDICAL LLC Tyrell Chemical HTN-130 AIR
Supplier:  TYRELL MEDICAL LLC HTN1305KGAIR
Encompass_Preferred
Inert dielectric high performance HFE thermal transfer fluid, 20kg/drum Air freight. Heat condution of machinery and equipment, electronics and datacenter cooling, immersion computer cooling. CAS 29770-93-9. 130C Boil Point
Catalog No. 50-273-4060