Learn More
Boston Bioproducts Inc Tyrode's Solution (with HEPES, without Calcium) - 1L
Supplier:  Boston Bioproducts Inc BSS3751L
Encompass
Tyrode’s Modified II is an isotonic buffer solution that plays a crucial role in cell culture and tissue preservation. It is formulated with inorganic salts, HEPES, and a carbohydrate energy source to provide cells with essential nutrients while maintaining a stable pH and osmotic balance, which is vital for optimal cell growth and functionality.The balanced composition of TBSS maintains the structural and physiological integrity of cells.
Catalog No. NC9041478
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.