Learn More
Sigma Aldrich Fine Chemicals Biosciences Tyrodes Salts Without sodi1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T214510X1L
Encompass
Tyrodes Salts play an important role in tissue culture. A salt SOLUTIONons basic function is to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions.
Catalog No. 50-290-1181