Learn More
TARGETMOL CHEMICALS INC TYROSINASE-IN-2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T602785MG
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tyrosinase-IN-2 (compound 67) is a potent tyrosinase inhibitor. Tyrosinase is a copper-containing metalloenzyme that is responsible for the rate-limiting catalytic step in the melanin biosynthesis and enzymatic browning. Tyrosinase-IN-2 has the potential for the research of skin whitening agents and food preservatives [1]. purity: 98%
Catalog No. 50-257-9629