Learn More
Medchemexpress LLC Tyrosinase-IN-7 | 2873418-52-9 | C15H10O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY151977100MG
Encompass_Preferred
Tyrosinase-IN-7 is a potent inhibitor of tyrosinase with an IC50 value of 1.57 μM. It effectively inhibits cell growth with low cytotoxicity, making it suitable for various research applications.
  • Potent tyrosinase inhibitor with an IC50 value of 1.57 μM
  • Inhibits cell growth with low cytotoxicity
  • Shows cytotoxicity with an IC50 value of 32 μM in MNT-1 whole cells
  • Exhibits a CC50 value of 108 μM against human MNT-1 cells
Catalog No. 50-003-73680