Learn More
Medchemexpress LLC Tyrosinase-related protein 2 (TRP-2) (181-188) | 187671-49-4 | 99.06% | 1088.25 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP252710MG
Encompass_Preferred
Tyrosinase-related Protein 2 (TRP-2) (181-188) is a tyrosinase-related protein 2 (TRP-2)-derived peptide, corresponding to residues 180-188. It is the major reactive epitope within TRP-2 recognized by anti-B16 CTLs. This peptide conforms to the MHC class I H2-Kb binding motif.
  • Derived peptide corresponding to residues 180-188
  • Major reactive epitope within TRP-2
  • Recognized by anti-B16 CTLs
  • Conforms to MHC class I H2-Kb binding motif
Catalog No. 50-004-23310