Learn More
Medchemexpress LLC Tyrosine Hydroxylase 10ug
SDP
Supplier:  Medchemexpress LLC HYP7356310UG
Encompass_Preferred
Tyrosine hydroxylase protein is a key catalyst in the biosynthesis of catecholamines including dopamine norepinephrine and epinephrine and oversees the conversion of L-tyrosine to L-dihydroxyphenylalanine (L-Dopa) which is the critical and rate-limiting step pathway in synthesis Tyrosine Hydroxylase Protein Mouse (sf9 His) is the recombinant mouse-derived Tyrosine Hydroxylase protein expressed by Sf9 insect cells with N-His labeled tag
Catalog No. 50-001-80084