Learn More
Medchemexpress LLC Tyrosine Hydroxylase Antibody (YA5393) | 59 kDa | 100 UL
Supplier: Medchemexpress LLC HYP85701100UL
Tyrosine Hydroxylase Antibody (YA5393) is a Mouse-derived and non-conjugated IgG1 monoclonal antibody, specifically designed to target Tyrosine Hydroxylase. It is purified using Protein G and is suitable for research applications such as immunohistochemistry (IHC) and immunofluorescence (IF).
- Mouse-derived IgG1 monoclonal antibody
- Non-conjugated
- Targets Tyrosine Hydroxylase
- Suitable for IHC and IF applications
- Reactivity with human, mouse, and rat samples
- Purified using Protein G