Learn More
Sino Biological Peptide Substrates: Tyrosine Kinase Substrate-3
Supplier:  Sino Biological T7158
Encompass_Preferred
The synthetic peptide Tyrosine Kinase Substrate-3 (RRLIEDAEYAARG) is derived from human SRC (amino acid 412-422) and considered to be a generic substrate for various protein tyrosine kinases.
Catalog No. 50-256-7582