Learn More
Medchemexpress LLC HY-N0474 100mg Medchemexpress, Tyrosol CAS:501-94-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0474100MG
Encompass_Preferred
Medchemexpress, HY-N0474 100mg Tyrosol CAS:501-94-0 Tyrosol is a derivative of phenethyl alcohol. Tyrosol attenuates pro-inflammatory cytokines from cultured astrocytes and NF-κB activation. Anti-oxidative and anti-inflammatory effects [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3589