Learn More
Cayman Chemical TyrothrIcIn 10mg
SDP
Supplier:  Cayman Chemical 2217710 MG
Encompass_Preferred
A polypeptide antibiotic mixture produced by B. brevis, composed of tyrocidine and gramicidin with rapid in vitro bactericidal activity against T. microdentium, T. macrodentium, T. vincentii, T. buccalis, T. mucosum, T. vaginalis, T. cruzi, and L. tropica; i.v. use leads to hemolysis and lethality (LD50s = 0.4 and 0.3 mg/kg, in dogs and mice, respectively)
Catalog No. 50-298-5694