Learn More
Enzo Life Sciences Tyrphostin 1 (100mg). CAS: 2826-26-8
SDP
Supplier:  Enzo Life Sciences BMLEI1900100
Encompass_Preferred
An inactive tyrphostin, which may be used as a negative control. Purity: ≥99%. Solubility: Soluble in 100% ethanol (25mg/ml, warm) or DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0598