Learn More
Enzo Life Sciences Tyrphostin 51 (5mg). CAS: 122520-90-5
SDP
Supplier:  Enzo Life Sciences BMLEI1890005
Encompass_Preferred
A potent inhibitor of EGF receptor kinase activity (IC50=0.8 µM). Inhibition is mixed competitive with ATP and substrate. Purity: ≥99%. Solubility: Slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0596