Learn More
Medchemexpress LLC Tyrphostin A51 | 126433-07-6 | MFCD00133903 | 99.0% | 268.23 g·mol⁻¹ | C13H8N4O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY101960A1MG
Encompass_Preferred
Tyrphostin A51 (CAS 126433-07-6) is a small-molecule protein tyrosine kinase inhibitor used in biochemical and cellular research to probe EGFR-related signaling and kinase-dependent processes. It is supplied as a solid with high reported purity and recommended storage conditions for short- and long-term stability.
  • Potent protein tyrosine kinase inhibitor for research use.
  • Useful for studying EGFR-associated signaling pathways.
  • High purity suitable for biochemical and cell-based assays.
  • Solid form with defined storage recommendations to preserve stability.
  • Available in small research quantities for assay development.
Catalog No. 50-002-47229