Learn More
Medchemexpress LLC Tyrphostin AG 1288 | 116313-73-6 | MFCD00236451 | 98.7% | 231.16 g/mol | C10H5N3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101961100MG
Encompass_Preferred
Tyrphostin AG 1288 is a small-molecule tyrosine kinase inhibitor used for research to probe kinase-mediated signaling. It inhibits ICAM1 expression and TNFα-mediated cytotoxicity in vitro and is supplied as a light yellow solid with high purity for biochemical and cell-based studies.
  • Potent tyrosine kinase inhibitor for research applications.
  • Reported to inhibit ICAM1 expression and TNFα-mediated cytotoxicity in vitro.
  • High purity (~98.7%) suitable for analytical and biological studies.
  • Molecular weight 231.16 g/mol; formula C10H5N3O4.
  • Stable as a powder with recommended cold storage for long-term integrity.
  • Available in multiple milligram pack sizes for assay flexibility.
Catalog No. 50-002-48682