Learn More
Medchemexpress LLC Tyrphostin AG 528 | 133550-49-9 | MFCD01076474 | >97.0% | 306.32 | C18H14N2O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10049950MG
Encompass_Preferred
Tyrphostin AG 528 is a small-molecule protein tyrosine kinase inhibitor that acts on epidermal growth factor receptor (EGFR) and ErbB2. Supplied as a powder for research use, it has reported IC50 values of 4.9 μM for EGFR and 2.1 μM for ErbB2; chemical identifiers include CAS 133550-49-9, formula C18H14N2O3, and molecular weight 306.32.
  • Targets EGFR and ErbB2 tyrosine kinases.
  • Reported IC50s: 4.9 μM (EGFR), 2.1 μM (ErbB2).
  • Provided as a powder with defined storage stability.
  • Chemical identifiers: CAS 133550-49-9 and formula C18H14N2O3.
  • Intended for research use only.
Catalog No. 50-002-14939