Learn More
Medchemexpress LLC Tyrphostin AG1296 | 146535-11-7 | MFCD00270913 | 99.69% | 266.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13894100MG
Encompass_Preferred
Tyrphostin AG1296 is a potent and selective inhibitor of platelet-derived growth factor receptor (PDGFR), inhibiting signaling of human PDGF α- and β-receptors, and the stem cell factor receptor (c-Kit). It is also an inhibitor of FLT3. It has a purity of 99.69%.
  • Potent and selective inhibitor of PDGFR
  • Inhibits human PDGF α- and β-receptors
  • Inhibits stem cell factor receptor (c-Kit)
  • Potent inhibitor of FLT3
  • Suppresses viability of PLX4032-resistant melanoma cells
  • Induces apoptosis of A375R cells
  • Inhibits PDGFR phosphorylation
  • Inhibits migration of A375R cells
  • Suppresses A375R tumor growth in nud/nud mice
  • Inhibits atherosclerotic plaque progression
Catalog No. 50-003-50333