Learn More
Medchemexpress LLC Tyrphostin AG30 | 122520-79-0 | MFCD09739079 | 99.7% | 205.17 g·mol⁻1 | C10H7NO4 | 100 MG

Supplier: Medchemexpress LLC HY118532100MG
Tyrphostin AG30 is a small-molecule inhibitor of epidermal growth factor receptor (EGFR) tyrosine kinase used in research to probe EGFR/c-ErbB signaling and STAT5 activation. It is supplied as a light yellow solid with high chemical purity and defined storage recommendations for powder and solutions.
- High purity (99.7%), for consistent experimental results.
- Molecular weight 205.17 g·mol⁻1 and formula C10H7NO4.
- Potent and selective inhibitor of EGFR tyrosine kinase.
- Solid, light yellow to yellow appearance, suitable for handling and formulation.
- Recommended storage: powder -20°C (long-term) or 4°C (short-term); solutions -80°C (long-term).
- Provided as a 100 mg research reagent for laboratory use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.