Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tyrphostin AG30 | 122520-79-0 | MFCD09739079 | 99.7% | 205.17 g/mol | C10H7NO4 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11853250MG

Encompass_Preferred

Tyrphostin AG30 is a small-molecule inhibitor of the epidermal growth factor receptor (EGFR) tyrosine kinase used for biochemical and cell-based research. It is supplied as a solid powder and as a DMSO solution, and has confirmed molecular properties and high chemical purity.

  • High purity: 99.7%
  • Molecular formula: C10H7NO4
  • Molecular weight: 205.17 g/mol
  • Formats: solid powder; 10 mM solution in DMSO (1 mL)
  • Intended for research use only

Catalog No. 50-003-07708


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.