Learn More
Medchemexpress LLC (+)-Tyrphostin B44 | 133550-37-5 | MFCD00209855 | 97.0% | 308.33 g/mol | C18H16N2O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101007A100MG
Encompass_Preferred
(+)-Tyrphostin B44 is a small-molecule EGFR inhibitor with reported antitumor activity, supplied as a light yellow to yellow solid with molecular formula C18H16N2O3 and high purity for research use. The compound is soluble in DMSO and manufacturer guidance provides recommended in vivo formulation protocols and storage conditions for maintaining stability.
  • Acts as an EGFR inhibitor with antitumor activity.
  • High purity suitable for research applications.
  • Light yellow to yellow solid appearance.
  • Soluble in DMSO at concentrations ≥100 mg/mL for in vitro use.
  • Includes recommended in vivo formulation protocols for administration.
  • Requires cold storage to maintain stability.
Catalog No. 50-002-11522