Learn More
Medchemexpress LLC Tz-thalidomide | 2087490-42-2 | 98.6% | 571.58 | C29H29N7O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10146010MG
Encompass_Preferred
Tz-Thalidomide is a tetrazine-tagged derivative of thalidomide used in degrader and click-chemistry research. It serves as an E3 ligase ligand and contains a tetrazine group capable of undergoing inverse electron-demand Diels-Alder (iEDDA) reactions with trans-cyclooctene (TCO) partners. Reported biochemical data indicate binding affinity for BRD4. Typical purity is 98.6% and the compound is supplied as a solid and as a 10 mM solution in DMSO.
Catalog No. 50-002-49175