Learn More
TA Instruments Tzero Press and Die Sets Kit, Includes Tzero Press Four Die Sets and One Box Each of Tzero Pans and Lids, Provides Performance Convenience in Crimp and Hermetic Encapsulation
Supplier:  TA Instruments 901600.901
Encompass_Preferred
Waters TA Instruments Tzero Press and Die Sets Kit includes the Tzero Press, four die sets, and one box each of Tzero Pans and Tzero Lids. It provides performance and convenience in crimp and hermetic encapsulation of a wide variety of materials. Die sets are color coded, magnetically attached, and tool free for operational simplicity. Designed for compatibility with Tzero and Standard Series pans and lids, it ensures consistent and accurate encapsulation.
Catalog No. 50-005-80878