Learn More
Selleck Chemical LLC U-104 S2866-1g
SDP
Supplier:  Selleck Chemical LLC S28661G
Encompass_Preferred
U-104 (MST-104 NSC 213841 SLC-0111 WBI-5111) is a potent carbonic anhydrase (CA) inhibitor for CA IX and CA XII with Ki of 45 1 nM and 4 5 nM respectively very low inhibition for CA I and CA II
Catalog No. 50-311-2437