Learn More
Abcam U-51605, PGI2 and T x A2 synthase inhibitor, 100UG
Supplier:  Abcam AB120908100UG
Encompass_Preferred
MW 332.49 g/mol, Purity >98%. Selective inhibitor of PGI₂ synthase (IC₅₀ = 2 μM) and TXA₂ synthase. PGH₂ analog. Partial agonist at TP receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7138