Learn More
Abcam U-69593, kappa1-opioid receptor agonist, 5MG
Supplier:  Abcam AB1417035MG
Encompass_Preferred
MW 356.5 Da, Purity >99%. Potent, selective κ1-opioid receptor agonist (EC50 = 80-109 nM). Active *in vivo*. Antinociceptive.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3352