Learn More
AdipoGen U-73122
SDP
Supplier:  AdipoGen AGCR13698M005
Encompass_Preferred
Chemical. CAS 112648-68-7. Formula C29H40N2O3. MW 464.6. Gasdermin D N-terminal fragment GSDMD-N-induced pyroptosis inhibitor. Protects against GSDMD-N cytotoxicity in macrophages or against lethal infection in mice. Useful agent for inflammasome research. Potent cell permeable phospholipase C PLC , phospholipase A2 and 5-LO 5-lipoxygenase inhibitor. Affects the phospholipases by inhibiting the hydrolysis of PPI phosphatidylinositol to IP3 inositol triphosphate, which in turn leads to a drop in cytosolic Ca2. Potent inhibitor of the SERCA pump in smooth muscle. Potently inhibits Ca2-release from isolated smooth muscle cells independent of its effect on PLC. Inhibits agonist-induced platelet aggregation. Potently inhibits human polymorphonuclear neutrophil adhesion on biological surfaces and exhibits antinociceptive activity in vivo. Shown to inhibit agonist-induced down-regulation of muscarinic receptors in SK-N-SH neuroblastoma cells.
Catalog No. 50-256-2205