Learn More
Medchemexpress LLC HY-108630 5mg Medchemexpress, U-73343 CAS:142878-12-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086305MG
Encompass_Preferred
Medchemexpress, HY-108630 5mg U-73343 CAS:142878-12-4 U-73343, an inhibitor of PLC (putative phospholipase C) -dependent processes, is an analog of U-73122 and can be used as a negative control [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2324