Learn More
Carbosynth LLC. µ-Conotoxin GIIIB (0.5 mg vial), 1ea, Disulfide bonds: (Disulfide bonds between Cys3-Cys15, Cys4-Cys20, and Cys10-Cys21), MW: 2640.2, Molecular formula: C101H175N39O30S7
SDP
Supplier:  Carbosynth LLC. PCN4217V0.5MG
Encompass_Preferred
Synonyms: Arg-Asp-Cys-Cys-Thr-Hyp-Hyp-Arg-Lys-Cys-Lys-Asp-Arg-Arg-Cys-Lys-Hyp-Met-Lys-Cys-Cys-Ala-NH2, Application: Na+ Channel Blocker: Specific for Skeletal Muscle, Storage Temperature: 2-10°C, References: S. Sato, H. Nakamura, Y. Ohizumi, J. Kobayashi, and Y. Hirata, FEBS Lett., 155, 277 (1983). (Original), L.J. Cruz, W.R. Gray, B.M. Olivera, R.D. Zeikus, L. Kerr, D. Yoshikami, and E. Moczydlowski, J. Biol. Chem., 260, 9280 (1985). (Naming), Y. Ohizumi, H. Nakamura, J. Kobayashi, and W.A. Catterall, J. Biol. Chem., 261, 6149 (1986). (Pharmacol.)
Catalog No. 50-168-6379
 Restricted Item