Learn More
Meso Scale Discovery U-PLEX Chemokine Combo 1 (mouse), Multiplex Immunoassay Kit, 96-well plate, detects 8 analytes including IP-10, KC/GRO, MCP-1, low LLOD 0.1 pg/mL, high ULOD 30,800 pg/mL
SDP
Supplier:  Meso Scale Discovery K15321K1
Encompass
The MESO SCALE DISCOVERY U-PLEX Chemokine Combo 1 is a multiplex immunoassay kit designed for the quantitative measurement of eight key mouse chemokines. Utilizing advanced MULTI-ARRAY technology, this kit provides exceptional sensitivity and a broad dynamic range for biomarker analysis, particularly in immunology and inflammation studies. It allows simultaneous detection of multiple analytes from small-volume samples, enhancing efficiency in research applications.
  • Multiplex immunoassay kit for measuring key mouse chemokines
  • Detects IP-10, KC/GRO, MCP-1, MDC, MIP-1α, MIP-1β, MIP-2, MIP-3α
  • Supported by low detection limits in the sub- to low pg/mL range
  • Compatible with various MESO SCALE DISCOVERY instruments
  • Utilizes electrochemiluminescence (ECL) technology for detection
  • Research use only
  • Suitable for EDTA plasma, serum, and cell culture supernatants
Catalog No. NC2831530