Learn More
Meso Scale Discovery U-PLEX Cytokine Combo 1 hu
SDP
Supplier:  Meso Scale Discovery K15045K1
Encompass
The Meso Scale Discovery U-PLEX Cytokine Combo 1 Human Kit is a multiplex immunoassay that simultaneously measures 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-10 IL-12p70 IL-13 IFN- GM-CSF TNF- and MCP-1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescent detection technology on patterned arrays in a quick 3 5 hour protocol The kit includes a 10-plex human cytokine standard and all necessary reagents and plates for 96 tests This multiplex assay allows high-throughput quantification of multiple cytokines from a small sample volume
Catalog No. 50-000-78274