Learn More
Meso Scale Discovery U-PLEX Human A2M ASYQP 25 PL
SDP
Supplier:  Meso Scale Discovery K151Q9K24
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X Read Buffer 10X Wash Buffer and Plate Seals This assay allows for rapid reproducible quantitation of A2M levels with a typical 4 5 hour protocol
Catalog No. 50-000-80587
Add to Cart
Request a Quote