Learn More
Meso Scale Discovery U-PLEX Human A2M ASYQP
SDP
Supplier:  Meso Scale Discovery K151Q9K21
Encompass
The Meso Scale Discovery U-PLEX Human A2M Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of Alpha-2-Macroglobulin (A2M) in human serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluent This assay allows for efficient and reproducible quantification of the A2M biomarker
Catalog No. 50-000-81264