Learn More
Meso Scale Discovery U-PLEX Human B-NGF ASYQP
SDP
Supplier:  Meso Scale Discovery K1516DK22
Encompass
The U-PLEX Human Beta-NGF Assay (K1516DK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure beta nerve growth factor ( -NGF) levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary U-PLEX platform technology offering high sensitivity and specificity for -NGF detection The kit is intended for research use only and provides researchers with a reliable tool for studying neurotrophin signaling pathways and neurodegenerative processes in various sample types
Catalog No. 50-000-80610