Learn More
Meso Scale Discovery U-PLEX Human CA1 ASYQP
SDP
Supplier:  Meso Scale Discovery K151J9K21
Encompass
The Meso Scale Discovery U-PLEX Human CA1 Assay is a multiplex immunoassay kit for measuring CA1 in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls It monitors CA1 levels related to diseases and biological processes with a quick 3 5-hour protocol for efficient workflow
Catalog No. 50-000-79546