Learn More
Meso Scale Discovery U-PLEX Human CD20 ASYQP
SDP
Supplier:  Meso Scale Discovery K151F7K21
Encompass
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that measures CD20 levels in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and controls It allows simultaneous measurement of CD20 levels along with other biomarkers in the U-PLEX platform The kit has high specificity and low cross-reactivity It requires 50 L sample volume per well and the assay time is approximately 3 hours
Catalog No. 50-000-78534