Learn More
Meso Scale Discovery U-PLEX Human CD20 ASYQP
SDP
Supplier:  Meso Scale Discovery K151F7K22
Encompass
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrator diluent wash buffer read buffer and a vial of lyophilized calibrator that must be reconstituted before use The kit can perform up to 80 tests in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-82033