Learn More
Meso Scale Discovery U-PLEX Human CD276/B7-H3 ASYQP
SDP
Supplier:  Meso Scale Discovery K151E7K22
Encompass
The Meso Scale Discovery U-PLEX Human CD276 (B7-H3) Assay Kit is a multiplex immunoassay for measuring CD276 (B7-H3) in human serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and necessary reagents Results are read on Meso Scale Discovery instruments like SECTOR Imagers and QuickPlex enabling quantitative measurement of CD276 important in immune regulation
Catalog No. 50-000-81982