Learn More
Meso Scale Discovery U-PLEX Human ENA-78 ASY
SDP
Supplier:  Meso Scale Discovery K151VEK1
Encompass
The Meso Scale Discovery U-PLEX Human ENA 7 1 Assay Kit is a multiplex immunoassay that measures 8 autoantibodies associated with autoimmune diseases It uses electrochemiluminescence detection technology and provides quantitative results from 25 L of serum or plasma in under 3 hours The 8 analytes measured are Sm RNP SS-A SS-B Scl-70 Jo-1 Centromere B and Ribosomal P This assay kit offers high sensitivity requires minimal hands-on time and is suitable for use in clinical research and drug development
Catalog No. 50-000-81932