Learn More
Meso Scale Discovery U-PLEX Human EPO ASY 25 PL
SDP
Supplier:  Meso Scale Discovery K151VXK4
Encompass
The Meso Scale Discovery U-PLEX Human EPO Assay is an electrochemiluminescent immunoassay for the quantitative measurement of erythropoietin (EPO) in human serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of EPO with a lower limit of detection of 0 14 mIU/mL and an upper limit of quantification of 400 mIU/mL The assay can be run on MSD s SECTOR Imager 6000 for high throughput screening This quantitative immunoassay allows sensitive accurate and rapid measurement of EPO which can aid in the diagnosis of anemias and bone marrow disorders
Catalog No. 50-000-81728
Add to Cart
Request a Quote