Learn More
Meso Scale Discovery U-PLEX Human EPO ASY
SDP
Supplier:  Meso Scale Discovery K151VXK2
Encompass
The Meso Scale Discovery U-PLEX Human EPO Assay Kit is a multiplex immunoassay for measuring erythropoietin (EPO) in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less sample volume it quantifies EPO from 0 04 pg/mL to 1000 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and an MSD read buffer aiding in diagnosing anemia and blood disorders
Catalog No. 50-000-79541