Learn More
Meso Scale Discovery U-PLEX Human EPO ASY384-well
SDP
Supplier:  Meso Scale Discovery K251VXK2
Encompass
The Meso Scale Discovery U-PLEX Human EPO 384-Well Assay is an electrochemiluminescent immunoassay for measuring erythropoietin (EPO) in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in a 384-well format With a dynamic range up to 10 000 pg/mL and requiring only 25 L of sample it supports high-throughput screening and pharmacokinetic studies This sensitive EPO assay is ideal for biomarker research and drug development
Catalog No. 50-000-80618
Only null left
Add to Cart
Request a Quote