Learn More
Meso Scale Discovery U-PLEX Human EPO ASYQP
SDP
Supplier:  Meso Scale Discovery K151VXK21
Encompass
The U-PLEX Human EPO Assay (K151VXK) is a multiplex immunoassay kit designed to quantitatively measure human erythropoietin (EPO) levels in biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for EPO detection commonly used in research applications studying anemia kidney disease and hematopoiesis The kit provides reliable reproducible results for laboratory analysis of EPO concentrations in serum plasma or other sample types
Catalog No. 50-000-79542