Learn More
Meso Scale Discovery U-PLEX Human Fractalkine ASY
SDP
Supplier:  Meso Scale Discovery K151VCK1
Encompass
The Meso Scale Discovery U-PLEX Human Fractalkine Assay Kit measures human fractalkine in serum plasma cell culture supernatants and lysates It uses electrochemiluminescence (ECL) technology for high sensitivity and a broad dynamic range The kit includes a plate with capture antibodies ECL-tagged detection antibodies calibrators and controls Sensitivity is 0 04 pg/mL with a range up to 2500 pg/mL requiring 50 L per well and taking about 3 5 hours It aids in studying immune function and inflammation
Catalog No. 50-000-81468