Learn More
Meso Scale Discovery U-PLEX Human FSH ASYQP
SDP
Supplier:  Meso Scale Discovery K1516GK21
Encompass
The MSD U-PLEX Human FSH Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of follicle-stimulating hormone (FSH) in human biological samples This singleplex assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The assay is designed for research use and provides reliable reproducible results for FSH detection in serum plasma and other relevant sample types
Catalog No. 50-000-78881